Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ino80e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153580.1 | NP_705808.1 | 5 | 676 | ||
GenBank mRNA | AK135531.1 | - | 7 | 713 | ||
BC025527.1 | - | 6 | 1065 | |||
BC026475.1 | - | 5 | 676 | 4489168 | ||
BC055928.1 | - | 7 | 1311 | 3995231 |