Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arid4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081195.1 | NP_001074664.1 | 16 | 1734 | ||
GenBank mRNA | AK050594.1 | - | 15 | 1541 | ||
AK149171.1 | - | 6 | 794 | |||
BC058209.1 | - | 16 | 1651 | |||
BC157975.1 | - | 15 | 1536 | 9007297 | ||
BC158109.1 | - | 15 | 1536 | 9007390 |