Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173418.3 | NP_775594.1 | 3 | 1038 | ||
GenBank mRNA | AK044383.1 | - | 3 | 1039 | ||
AK044611.1 | - | 3 | 1040 | |||
BC037728.1 | - | 3 | 938 | |||
BC057628.1 | - | 3 | 1013 | 4511498 |