Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaper | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081341.1 | NP_001074810.1 | 31 | 4174 | ||
GenBank mRNA | AK048755.1 | - | 20 | 2702 | ||
AK173175.1 | - | 30 | 4104 | |||
BC052188.1 | - | 5 | 914 | |||
BC157959.1 | - | 30 | 4119 | 9007286 | ||
BC158062.1 | - | 30 | 4096 | 9088165 | ||
BC172119.1 | - | 30 | 4119 | 9054405 | ||
BC172151.1 | - | 30 | 4096 | 9054821 |