Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTN3A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006994.4 | NP_008925.1 | 5 | 703 | ||
NM_197974.2 | NP_932078.2 | 5 | 632 | |||
GenBank mRNA | AB209581.1 | - | 3 | 1973 | ||
AK223279.1 | - | 5 | 638 | |||
AK291722.1 | - | 5 | 687 | |||
AK301553.1 | - | 5 | 616 | |||
BC015815.2 | - | 5 | 640 | 3677284 | ||
BT007251.1 | - | 3 | 460 | |||
U90548.1 | - | 5 | 631 |