Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armcx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007578.2 | NP_001007579.1 | 2 | 159 | ||
GenBank mRNA | AK148507.1 | - | 2 | 158 | ||
AK160766.1 | - | 2 | 160 | |||
BC027264.1 | - | 2 | 152 | 3492516 | ||
BY777182.1 | - | 2 | 139 |