Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138584.2 | NP_613050.1 | 7 | 817 | ||
GenBank mRNA | AK009332.1 | - | 7 | 809 | ||
AK011985.1 | - | 7 | 784 | |||
AK090017.1 | - | 7 | 805 | |||
AK149930.1 | - | 7 | 791 | |||
AK150215.1 | - | 7 | 818 | |||
AK150581.1 | - | 7 | 795 | |||
AK153284.1 | - | 7 | 779 | |||
AK170666.1 | - | 7 | 810 | |||
AK187404.1 | - | 3 | 158 | |||
AK206667.1 | - | 3 | 157 | |||
AK209522.1 | - | 3 | 157 | |||
AK210536.1 | - | 3 | 157 | |||
AK211454.1 | - | 3 | 157 | |||
AK217468.1 | - | 3 | 157 | |||
AK218378.1 | - | 3 | 157 | |||
BC099434.1 | - | 6 | 582 | 30714184 | ||
BC132377.1 | - | 7 | 794 | 40130654 | ||
BC132403.1 | - | 7 | 794 | 40130680 |