Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138596.2 | NP_613062.1 | 3 | 267 | ||
GenBank mRNA | AK003862.1 | - | 3 | 261 | ||
AK013992.1 | - | 3 | 271 | |||
AK156498.1 | - | 3 | 276 | |||
AK215905.1 | - | 3 | 115 | |||
BC048638.1 | - | 3 | 260 | 6771841 | ||
BC056438.1 | - | 3 | 256 | 6818638 | ||
BC059231.1 | - | 2 | 2749 | |||
BC089312.1 | - | 2 | 1812 |