Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081132.1 | NP_001074601.1 | 15 | 3027 | ||
GenBank mRNA | BC039278.1 | - | 4 | 690 | ||
BC138358.1 | - | 15 | 2990 | 8861378 | ||
BC138359.1 | - | 15 | 2990 | 8861379 |