Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009092.3 | NP_033118.1 | 4 | 328 | ||
GenBank mRNA | AK007992.1 | - | 4 | 336 | ||
AK010647.1 | - | 4 | 330 | |||
AK012781.1 | - | 4 | 317 | |||
AK158902.1 | - | 3 | 394 | |||
AK162022.1 | - | 4 | 322 | |||
AK162335.1 | - | 4 | 320 | |||
AK167206.1 | - | 4 | 322 | |||
BC081466.1 | - | 3 | 315 | 6412006 | ||
BC086900.1 | - | 4 | 321 | 6760363 | ||
BC086901.1 | - | 4 | 321 | 6763239 | ||
D25213.1 | - | 3 | 309 |