Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039048.2 | NP_001034137.2 | 5 | 733 | ||
GenBank mRNA | BC147663.1 | - | 5 | 701 | 9087452 | |
BC147666.1 | - | 5 | 701 | 9087455 | ||
CK128385.1 | - | 3 | 276 | |||
DQ229108.1 | - | 5 | 629 |