Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARIH2 SIRNA Assays ›
Gene Symbol:
ARIH2
Gene Name:
ariadne RBR E3 ubiquitin protein ligase 2
Gene Aliases:
ARI2, TRIAD1
Chromosome Location:
Chr.3: 48918820 - 48985541 on Build GRCh38
UniGene ID:
Hs.633601
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s20391
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARIH2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001317333.1
NP_001304262.1
15
1742
NM_001317334.1
NP_001304263.1
12
1204
NM_006321.3
NP_006312.1
14
1620
XM_011533265.1
XP_011531567.1
14
1677
XM_011533266.1
XP_011531568.1
15
1799
XM_011533267.1
XP_011531569.1
14
1731
XM_011533268.1
XP_011531570.1
13
1624
XM_011533270.1
XP_011531572.1
15
1711
XM_011533271.1
XP_011531573.1
14
1361
XM_011533273.1
XP_011531575.1
16
1742
XM_011533275.2
XP_011531577.1
14
1562
XM_011533276.1
XP_011531578.1
9
1018
XM_017005531.1
XP_016861020.1
14
1662
XM_017005532.1
XP_016861021.1
13
1555
XM_017005533.1
XP_016861022.1
13
2071
XM_017005534.1
XP_016861023.1
13
1280
XM_017005535.1
XP_016861024.1
13
1488
XM_017005536.1
XP_016861025.1
16
1776
XM_017005537.1
XP_016861026.1
15
1642
XM_017005538.1
XP_016861027.1
15
1688
XM_017005539.1
XP_016861028.1
14
1292
XM_017005540.1
XP_016861029.1
16
1673
XM_017005541.1
XP_016861030.1
16
1743
GenBank mRNA
AB208857.1
-
8
755
AF099149.1
-
13
1424
AF183427.1
-
13
1426
AJ130978.1
-
13
1424
AK091240.1
-
13
2167
AK223550.1
-
15
1726
AK307694.1
-
11
970
AK313434.1
-
12
1284
BC000422.2
-
14
1580
2964505
CR456785.1
-
12
1280
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
multicellular organism development
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
developmental cell growth
protein K63-linked ubiquitination
protein K48-linked ubiquitination
hematopoietic stem cell proliferation
positive regulation of protein targeting to mitochondrion
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
ubiquitin conjugating enzyme binding
ubiquitin protein ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0