Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286260.1 | NP_001273189.1 | 5 | 1455 | ||
NM_018823.2 | NP_061293.2 | 6 | 1509 | |||
NM_133957.3 | NP_598718.2 | 7 | 1571 | |||
GenBank mRNA | AF162853.1 | - | 5 | 1366 | ||
AF200687.1 | - | 6 | 1144 | |||
AF228705.1 | - | 3 | 1209 | |||
AF228706.1 | - | 3 | 1209 | |||
AF369980.1 | - | 7 | 1210 | |||
AF453571.2 | - | 7 | 1179 | |||
AK134022.1 | - | 7 | 1575 |