Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018819.4 | NP_061289.1 | 5 | 653 | ||
GenBank mRNA | AF181116.1 | - | 5 | 609 | ||
AK002241.1 | - | 5 | 654 | |||
AK002266.1 | - | 5 | 632 | |||
AK004013.1 | - | 5 | 609 | |||
AK005348.1 | - | 5 | 645 | |||
AK013391.1 | - | 5 | 654 | |||
AK014421.1 | - | 5 | 611 | |||
AK014435.1 | - | 5 | 883 | |||
AK135943.1 | - | 5 | 624 | |||
AK170514.1 | - | 5 | 670 | |||
BC024365.1 | - | 5 | 615 | 4988147 | ||
BC103767.1 | - | 5 | 656 | 30840199 |