Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021303.2 | NP_067278.2 | 17 | 2083 | ||
GenBank mRNA | AF155546.1 | - | 5 | 593 | ||
AK076287.1 | - | 16 | 2125 | |||
AK133834.1 | - | 17 | 2083 | |||
AK157765.1 | - | 17 | 2076 | |||
AK199140.1 | - | 1 | 86 | |||
BC020013.1 | - | 17 | 2054 | 3663482 |