Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021328.3 | NP_067303.1 | 9 | 754 | ||
GenBank mRNA | AF244361.1 | - | 9 | 734 | ||
AF271733.1 | - | 9 | 797 | |||
AK005992.1 | - | 9 | 763 | |||
AK040685.1 | - | 9 | 754 | |||
AK170915.1 | - | 9 | 795 | |||
BC026543.1 | - | 9 | 739 | 5040094 |