Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207523.2 | NP_997406.1 | 5 | 509 | ||
GenBank mRNA | AK028387.1 | - | 5 | 482 | ||
AK152095.1 | - | 5 | 482 | |||
BC016558.1 | - | 4 | 448 | |||
BC026656.1 | - | 5 | 466 | |||
BC029892.1 | - | 5 | 468 | 4163533 | ||
BC086883.1 | - | 5 | 484 | 6756980 | ||
BC086884.1 | - | 5 | 482 | 30122733 | ||
BC096637.1 | - | 5 | 467 | |||
BC125640.1 | - | 5 | 488 | 40130155 | ||
BC125642.1 | - | 5 | 488 | 40130157 | ||
BY585987.1 | - | 4 | 350 |