Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm7073 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039240.3 | NP_001034329.2 | 4 | 805 | ||
GenBank mRNA | AK135660.1 | - | 4 | 807 |