Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006348.3 | NP_006339.3 | 22 | 3011 | ||
NM_181733.2 | NP_859422.2 | 21 | 2948 | |||
GenBank mRNA | AF058718.1 | - | 21 | 2474 | ||
AK022835.1 | - | 13 | 1387 | |||
AK123726.1 | - | 1 | 1344 | |||
AK314270.1 | - | 22 | 3009 |