Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem14c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025387.3 | NP_079663.1 | 6 | 792 | ||
GenBank mRNA | AK003891.1 | - | 6 | 741 | ||
AK008495.1 | - | 5 | 682 | |||
AK008878.1 | - | 6 | 714 | |||
AK010609.1 | - | 6 | 752 | |||
AK012101.1 | - | 6 | 739 | |||
AK019395.1 | - | 6 | 741 | |||
AK050688.1 | - | 6 | 727 | |||
AK089021.1 | - | 5 | 666 | |||
AK201301.1 | - | 1 | 164 | |||
BC025854.1 | - | 6 | 693 | 5250659 | ||
BC055862.1 | - | 6 | 719 | 3492090 | ||
BC069899.1 | - | 5 | 1438 |