Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025558.5 | NP_079834.2 | 1 | 186 | ||
GenBank mRNA | AK003333.2 | - | 1 | 187 | ||
AK003815.1 | - | 1 | 176 | |||
AK007780.1 | - | 206 | ||||
AK019263.1 | - | 1 | 186 | |||
AK052810.1 | - | 1 | 187 | |||
AK088970.1 | - | 1 | 188 | |||
AK165543.1 | - | 1 | 190 | |||
AK166544.1 | - | 1 | 164 | |||
AK168422.1 | - | 1 | 176 | |||
BC046479.1 | - | 1 | 149 | |||
BC046810.1 | - | 1 | 152 | |||
BC048147.1 | - | 1 | 176 | |||
BC054749.1 | - | 1 | 207 | 6410090 | ||
BC058812.1 | - | 1 | 149 | 6530835 | ||
BC062980.1 | - | 1 | 198 | 6842939 |