Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006360.5 | NP_006351.2 | 2 | 180 | ||
XM_011519840.2 | XP_011518142.1 | 2 | 204 | |||
GenBank mRNA | AF064603.1 | - | 2 | 110 | ||
AK131064.1 | - | 2 | 134 | |||
AK222564.1 | - | 2 | 99 | |||
AK292139.1 | - | 2 | 122 | |||
AK300002.1 | - | 2 | 124 | |||
AK307121.1 | - | 2 | 99 | |||
AK312512.1 | - | 2 | 149 | |||
AY769947.1 | - | 2 | 59 | |||
BC019103.2 | - | 2 | 102 | 5021401 | ||
BC051292.1 | - | 2 | 104 | 3607828 | ||
CR450300.1 | - | 2 | 59 | |||
DB479224.1 | - | 2 | 180 | |||
DQ185042.1 | - | 2 | 50 |