Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ormdl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025661.4 | NP_079937.1 | 3 | 432 | ||
GenBank mRNA | AK009621.1 | - | 3 | 433 | ||
AK012718.1 | - | 3 | 402 | |||
AK087497.1 | - | 3 | 401 | |||
AK154085.1 | - | 3 | 424 | |||
BC026412.1 | - | 1 | 107 | |||
BC046594.1 | - | 3 | 370 | 6308578 |