Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm8765 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244649.1 | NP_001231578.1 | 4 | 1326 | ||
GenBank mRNA | BC172081.1 | - | 4 | 1326 | 9054367 |