Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Use1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145780.1 | NP_001139252.1 | 9 | 1015 | ||
NM_025917.4 | NP_080193.1 | 8 | 802 | |||
GenBank mRNA | AF353245.1 | - | 8 | 795 | ||
AK003257.1 | - | 8 | 795 | |||
AK003385.1 | - | 7 | 666 | |||
AK008573.1 | - | 8 | 795 | |||
AK008661.1 | - | 7 | 700 | |||
AK012490.1 | - | 8 | 796 | |||
AK017484.1 | - | 5 | 869 | |||
AK160726.1 | - | 8 | 795 | |||
AK165639.1 | - | 8 | 944 | |||
AY842453.1 | - | 8 | 781 | |||
BC025086.1 | - | 7 | 878 | 3995001 | ||
BC038519.1 | - | 7 | 865 | 1428854 | ||
BC075695.1 | - | 9 | 921 | 30623689 |