Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025928.3 | NP_080204.1 | 4 | 382 | ||
GenBank mRNA | AF348509.1 | - | 2 | 185 | ||
AF348510.1 | - | 4 | 392 | |||
AK011174.1 | - | 4 | 384 | |||
AK012113.1 | - | 4 | 383 | |||
AK013787.1 | - | 4 | 385 | |||
AK078308.1 | - | 4 | 401 | |||
AK150603.1 | - | 4 | 399 | |||
AK168200.1 | - | 4 | 383 | |||
AK180171.1 | - | 2 | 117 | |||
BC034333.1 | - | 4 | 360 | |||
BC037139.1 | - | 4 | 391 | 4952550 | ||
BC049223.1 | - | 4 | 381 | 3257320 |