Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033136.3 | NP_001028308.1 | 8 | 1148 | ||
GenBank mRNA | AK004786.1 | - | 8 | 1074 | ||
AK146299.1 | - | 8 | 1067 | |||
AK151090.1 | - | 8 | 1069 | |||
AK153322.1 | - | 8 | 1069 | |||
AK153369.1 | - | 8 | 1068 | |||
AK171770.1 | - | 8 | 1149 | |||
BC055754.1 | - | 8 | 1042 |