Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024216.1 | NP_077178.1 | 4 | 547 | ||
GenBank mRNA | AK016996.1 | - | 4 | 478 | ||
AK042405.1 | - | 1170 | ||||
AK080720.1 | - | 4 | 494 | |||
AK087968.1 | - | 4 | 498 | |||
AK146783.1 | - | 4 | 485 | |||
BC003341.1 | - | 4 | 547 | 3257412 |