Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsmce2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026746.3 | NP_081022.2 | 7 | 845 | ||
GenBank mRNA | AK003685.1 | - | 7 | 797 | ||
BC009125.1 | - | 7 | 756 | 3155917 |