Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam25c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183278.2 | NP_899101.1 | 2, 3 | 171 | ||
GenBank mRNA | AK008614.1 | - | 2, 3 | 171 | ||
BC086894.1 | - | 2, 3 | 173 | |||
BC147104.1 | - | 2, 3 | 160 | 9056159 | ||
BC147105.1 | - | 2, 3 | 160 | 9056136 |