Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153173.4 | NP_694813.1 | 1 | 390 | ||
GenBank mRNA | AA028777.1 | - | 1 | 387 | ||
AK007642.1 | - | 1 | 386 | |||
BC028550.1 | - | 1 | 331 | 1398106 | ||
BC058529.1 | - | 1 | 327 | 6817839 | ||
BC087952.1 | - | 1 | 379 | 6787571 |