Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190310.1 | NP_001177239.1 | 2 | 207 | ||
NM_027218.3 | NP_081494.1 | 2 | 207 | |||
GenBank mRNA | AA914236.1 | - | 2 | 207 | ||
AK007794.1 | - | 2 | 164 | |||
AY230259.1 | - | 2 | 171 | |||
AY230260.1 | - | 2 | 171 | |||
AY365134.1 | - | 2 | 103 |