Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145929.1 | NP_001139401.1 | 3 | 625 | ||
NM_175152.4 | NP_780361.2 | 4 | 727 | |||
GenBank mRNA | AK008225.1 | - | 4 | 699 | ||
AK038184.1 | - | 4 | 701 | |||
AK050740.1 | - | 4 | 741 | |||
BC059091.1 | - | 3 | 610 | |||
BC094395.1 | - | 4 | 708 | 5700290 | ||
BC115485.1 | - | 4 | 726 | |||
BC115486.1 | - | 4 | 724 |