Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asnsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290984.1 | NP_001277913.1 | 5 | 514 | ||
NM_133728.3 | NP_598489.2 | 5 | 1945 | |||
GenBank mRNA | AK031295.1 | - | 5 | 1903 | ||
AK034746.1 | - | 1864 | ||||
AK049194.1 | - | 5 | 1892 | |||
AK050045.1 | - | 5 | 1885 | |||
AK076793.1 | - | 5 | 1847 | |||
AK155030.1 | - | 5 | 1884 | |||
AK168103.1 | - | 5 | 1885 | |||
AK168534.1 | - | 5 | 515 | |||
AK170510.1 | - | 5 | 1861 | |||
BC013617.1 | - | 5 | 1765 | 3979452 |