Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027439.4 | NP_081715.1 | 8 | 918 | ||
GenBank mRNA | AB192471.2 | - | 8 | 794 | ||
AK017482.1 | - | 8 | 850 | |||
AK029405.1 | - | 8 | 821 | |||
AK076495.1 | - | 8 | 863 | |||
AK151735.1 | - | 8 | 850 | |||
AK168139.1 | - | 8 | 875 | |||
AW985491.1 | - | 5 | 485 | |||
BC014706.1 | - | 8 | 789 | 4457079 |