Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ier5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030244.3 | NP_084520.3 | 1 | 1398 | ||
GenBank mRNA | AK012145.1 | - | 1 | 1411 | ||
BC003464.1 | - | 1 | 1397 | 2648900 | ||
BC010234.1 | - | 1 | 1398 | 3499467 | ||
BC040084.1 | - | 1 | 1398 | 3499467 |