Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004331.2 | NP_004322.1 | 3 | 441 | ||
XM_005273617.3 | XP_005273674.1 | 3 | 345 | |||
GenBank mRNA | AB004788.1 | - | 3 | 362 | ||
AF060922.1 | - | 3 | 395 | |||
AF067396.1 | - | 3 | 399 | |||
AF079221.1 | - | 3 | 399 | |||
AF452712.2 | - | 3 | 414 | |||
AK290213.1 | - | 3 | 357 | |||
AK300286.1 | - | 3 | 366 | |||
AK300727.1 | - | 3 | 380 | |||
AK315870.1 | - | 3 | 318 | |||
AK316315.1 | - | 3 | 345 | |||
AL132665.1 | - | 3 | 415 | |||
BC001559.1 | - | 3 | 341 | 3459327 | ||
BC009603.1 | - | 3 | 409 | 4134765 | ||
BT019500.1 | - | 3 | 317 | |||
BT019501.1 | - | 3 | 317 | |||
CD558918.1 | - | 3 | 550 | |||
CR456701.1 | - | 3 | 317 |