Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028500.3 | NP_082776.2 | 4 | 529 | ||
NM_029782.3 | NP_084058.2 | 2 | 230 | |||
GenBank mRNA | AB206786.1 | - | 4 | 434 | ||
AK006582.1 | - | 4 | 530 | |||
AK018263.1 | - | 2 | 152 | |||
AK046081.1 | - | 2 | 231 |