Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Whrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008791.2 | NP_001008791.1 | 9 | 2432 | ||
NM_001008792.2 | NP_001008792.1 | 8 | 2411 | |||
NM_001008793.2 | NP_001008793.1 | 8 | 2399 | |||
NM_028640.2 | NP_082916.1 | 8 | 2399 | |||
GenBank mRNA | AK122523.1 | - | 8 | 1530 | ||
AK190484.1 | - | 4 | 310 | |||
AY739114.1 | - | 8 | 2359 | |||
AY739115.1 | - | 9 | 2392 | |||
AY739116.1 | - | 8 | 2371 | |||
AY739117.1 | - | 8 | 2359 | |||
AY739118.1 | - | 5 | 1008 | |||
AY739119.1 | - | 4 | 975 | |||
AY739120.1 | - | 3 | 762 | |||
AY739121.1 | - | 3 | 880 | |||
AY739122.1 | - | 4 | 1212 | |||
BC150757.1 | - | 8 | 1756 | 9087668 |