Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dynap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029346.1 | NP_083622.1 | 3 | 541 | ||
GenBank mRNA | AK009098.1 | - | 3 | 356 | ||
HQ404309.1 | - | 3 | 454 | |||
HQ404310.1 | - | 3 | 454 | |||
HQ404311.1 | - | 3 | 454 |