Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029751.4 | NP_084027.1 | 5 | 890 | ||
GenBank mRNA | AK161942.1 | - | 5 | 860 |