Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006404.4 | NP_006395.2 | 2 | 420 | ||
XM_011528496.1 | XP_011526798.1 | 2 | 263 | |||
GenBank mRNA | AK314887.1 | - | 2 | 386 | ||
BC014451.1 | - | 2 | 284 | 4907433 | ||
BG423815.1 | - | 2 | 386 | |||
BM808358.1 | - | 2 | 438 | |||
CR456948.1 | - | 2 | 202 | |||
HY110044.1 | - | 2 | 419 | |||
L35545.1 | - | 2 | 226 | |||
X89079.1 | - | 2 | 231 |