Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Clip4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271483.1 | NP_001258412.1 | 3 | 409 | ||
NM_001271484.1 | NP_001258413.1 | 3 | 494 | |||
NM_030179.3 | NP_084455.2 | 3 | 397 | |||
NM_175378.1 | NP_780587.1 | 3 | 494 | |||
GenBank mRNA | AK018895.1 | - | 3 | 487 | ||
AK019516.1 | - | 3 | 410 | |||
AK029962.1 | - | 3 | 495 | |||
AK075758.1 | - | 3 | 485 | |||
AK161301.1 | - | 3 | 484 | |||
BC035226.1 | - | 3 | 397 | 4920788 |