Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053177.1 | NP_444407.1 | 10 | 1300 | ||
GenBank mRNA | AF302009.1 | - | 10 | 1297 | ||
AK028385.1 | - | 10 | 1277 | |||
AK034471.1 | - | 10 | 1311 | |||
AK038594.1 | - | 10 | 1313 | |||
AK049606.1 | - | 10 | 1301 | |||
BC005651.1 | - | 10 | 1300 | 3257440 | ||
BC020377.1 | - | 6 | 1742 |