Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026122.4 | NP_080398.1 | 2 | 210 | ||
NM_175074.2 | NP_778249.1 | 2 | 210 | |||
GenBank mRNA | AK002970.1 | - | 2 | 129 | ||
AK003272.1 | - | 2 | 158 | |||
AK153223.1 | - | 2 | 170 | |||
AK214566.1 | - | 2 | 58 | |||
BC005693.1 | - | 2 | 146 | 3597594 | ||
CJ150650.1 | - | 2 | 211 |