Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302758.1 | NP_001289687.1 | 4 | 423 | ||
NM_006433.4 | NP_006424.2 | 3 | 342 | |||
NM_012483.3 | NP_036615.2 | 4 | 586 | |||
XM_005264084.2 | XP_005264141.1 | 4 | 356 | |||
XM_005264085.2 | XP_005264142.1 | 3 | 331 | |||
XM_005264087.2 | XP_005264144.1 | 4 | 583 | |||
GenBank mRNA | AK304728.1 | - | 4 | 356 | ||
AK308781.1 | - | 3 | 445 | |||
AK310057.1 | - | 4 | 584 | |||
AK311012.1 | - | 3 | 295 | |||
AK311537.1 | - | 4 | 476 | |||
AK311538.1 | - | 3 | 209 | |||
AK311657.1 | - | 3 | 872 | |||
BC023576.2 | - | 3 | 217 | 4766109 | ||
CR541859.1 | - | 3 | 214 | |||
X05044.1 | - | 4 | 446 | |||
X54101.1 | - | 3 | 342 |