Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001248006.1 | NP_001234935.1 | 6 | 1554 | ||
NM_001248007.1 | NP_001234936.1 | 5 | 1217 | |||
NM_006458.3 | NP_006449.2 | 7 | 1555 | |||
NM_033278.3 | NP_150594.2 | 6 | 1390 | |||
XM_011519844.1 | XP_011518146.1 | 7 | 1654 | |||
XM_011519845.2 | XP_011518147.1 | 6 | 1335 | |||
XM_017017098.1 | XP_016872587.1 | 6 | 1366 | |||
XM_017017099.1 | XP_016872588.1 | 6 | 1258 | |||
GenBank mRNA | AF045239.1 | - | 5 | 1159 | ||
AF220020.1 | - | 5 | 1159 | |||
AF220021.1 | - | 6 | 1126 | |||
AF220022.1 | - | 5 | 922 | |||
AK126685.1 | - | 4 | 783 | |||
AK291396.1 | - | 6 | 1554 | |||
AK298843.1 | - | 5 | 1093 | |||
AK299586.1 | - | 6 | 1347 | |||
AK311127.1 | - | 6 | 1345 | |||
AL360185.1 | - | 1 | 45 | |||
AL360192.1 | - | 6 | 1096 | |||
BC096827.1 | - | 6 | 1390 | 6108991 | ||
DA347370.1 | - | 1 | 420 |