Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319674.1 | NP_001306603.1 | 3 | 308 | ||
NM_006582.3 | NP_006573.2 | 3 | 338 | |||
NM_024482.2 | NP_077808.1 | 3 | 305 | |||
XM_011540518.2 | XP_011538820.1 | 3 | 441 | |||
XM_011540519.2 | XP_011538821.1 | 3 | 281 | |||
XM_011540521.2 | XP_011538823.1 | 3 | 410 | |||
XM_011540522.2 | XP_011538824.1 | 3 | 250 | |||
XM_017000087.1 | XP_016855576.1 | 3 | 456 | |||
GenBank mRNA | AF099013.1 | - | 2 | 202 | ||
AF173868.1 | - | 3 | 233 | |||
AF203694.1 | - | 3 | 232 | |||
AK000892.1 | - | 3 | 304 | |||
AK023126.1 | - | 3 | 225 | |||
AK315195.1 | - | 3 | 284 | |||
AK315436.1 | - | 3 | 242 | |||
BC001473.2 | - | 3 | 255 | 3138481 | ||
DA919003.1 | - | 3 | 335 |