Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930427A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134041.3 | NP_598802.2 | 8 | 2045 | ||
GenBank mRNA | AK029651.1 | - | 8 | 2047 | ||
AK167092.1 | - | 8 | 2004 | |||
AK220305.1 | - | 2133 | ||||
BC116830.1 | - | 4 | 1384 | |||
BC116834.1 | - | 4 | 1384 |