Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013674.1 | NP_038702.1 | 7 | 1118 | ||
GenBank mRNA | AK089319.1 | - | 7 | 1137 | ||
AK089819.1 | - | 7 | 1140 | |||
AK154138.1 | - | 7 | 1142 | |||
AK154315.1 | - | 7 | 1130 | |||
AK156562.1 | - | 7 | 1140 | |||
BC137713.1 | - | 7 | 1106 | 8860732 | ||
BC137714.1 | - | 7 | 1106 | 8860733 | ||
U11692.1 | - | 7 | 1118 | |||
U34307.1 | - | 7 | 1112 |